1-(2-Chloroethyl)-3-(3-fluorophenyl)urea

Lieferant: Apollo Scientific
Achtung

PC46359-1G PC46359-250MG
APOSPC46359-1GEA 26.1 EUR
APOSPC46359-1G APOSPC46359-250MG
1-(2-Chloroethyl)-3-(3-fluorophenyl)urea
1-(2-Chloroethyl)-3-(3-fluorophenyl)urea
Formel: C₉H₁₀ClFN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD14586691
CAS-Nummer: 146257-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR