N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide 97%
Lieferant: Apollo Scientific
PC4636-1G
PC4636-500MG
PC4636-5G
APOSPC4636-1GEA
172
EUR
APOSPC4636-1G
APOSPC4636-500MG
APOSPC4636-5G
N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide 97%
N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide
Formel:
C₁₃H₁₂ClF₆NO Molecular Weight: 347.69 g/mol Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793437 CAS-Nummer: 478261-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...