1-(2-Bromethoxy)-3-(trifluormethyl)benzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(2-Bromoethoxy)benzotrifluoride
PC4638-1G
APOSPC4638-1GEA
21.9
EUR
APOSPC4638-1G
1-(2-Bromethoxy)-3-(trifluormethyl)benzol
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene
Formel:
C₉H₈BrF₃O Molecular Weight: 269,06 g/mol Siedepunkt: 116…118 °C/10mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215501 CAS-Nummer: 18800-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...