2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

PC4646-1G PC4646-500MG
APOSPC4646-1GEA 200 EUR
APOSPC4646-1G APOSPC4646-500MG
2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine 97%
2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine
Formel: C₁₅H₂₀F₃N₅
Molecular Weight: 327.348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00728816
CAS-Nummer: 516494-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR