2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4646-1G
PC4646-500MG
APOSPC4646-1GEA
200
EUR
APOSPC4646-1G
APOSPC4646-500MG
2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine 97%
2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine
Formel:
C₁₅H₂₀F₃N₅ Molecular Weight: 327.348 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00728816 CAS-Nummer: 516494-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...