2-(Piperidin-2-ylmethyl)-3,3,3-trifluoropropanoic acid

Lieferant: Apollo Scientific
Achtung

PC4651-250MG
APOSPC4651-250MGEA 105 EUR
APOSPC4651-250MG
2-(Piperidin-2-ylmethyl)-3,3,3-trifluoropropanoic acid
2-(Piperidin-2-ylmethyl)-3,3,3-trifluoropropanoic acid
Formel: C₉H₁₄F₃NO₂
Molecular Weight: 225,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427205
CAS-Nummer: 480438-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR