1,1,1,3,3,3-Hexafluoroisopropyl-2-fluoroacrylate 97%

Lieferant: Apollo Scientific
Achtung

PC4746-1G
APOSPC4746-1GEA 328 EUR
APOSPC4746-1G
1,1,1,3,3,3-Hexafluoroisopropyl-2-fluoroacrylate 97%
1,1,1,3,3,3-Hexafluoroisopropyl-2-fluoroacrylate
Formel: C₆H₃F₇O₂
Molecular Weight: 240,08 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03094251
CAS-Nummer: 74359-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR