2,4-Difluoro-3-(trifluoromethyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

PC47684-1G PC47684-250MG
APOSPC47684-1GEA 532 EUR
APOSPC47684-1G APOSPC47684-250MG
2,4-Difluoro-3-(trifluoromethyl)aniline 95%
2,4-Difluoro-3-(trifluoromethyl)aniline
Formel: C₇H₄F₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD09261074
CAS-Nummer: 123973-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR