1H,1H,2H,3H,3H-Perfluoroheptane-1,2-diol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,5,5,6,6,7,7,7-Nonafluoroheptane-1,2-diol

PC4785-1G
APOSPC4785-1GEA 75.4 EUR
APOSPC4785-1G
1H,1H,2H,3H,3H-Perfluoroheptane-1,2-diol 98%
1H,1H,2H,3H,3H-Perfluoroheptane-1,2-diol
Formel: C₇H₇F₉O₂
Molecular Weight: 294,115 g/mol
Siedepunkt: 104…106 °C (5 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03424478
CAS-Nummer: 125070-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR