Hexafluorokieselsäure 23% in wässriger Lösung
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Fluosilicic acid
PC4797-1KG
APOSPC4797-1KGEA
203
EUR
APOSPC4797-1KG
Hexafluorokieselsäure 23% in wässriger Lösung
Hexafluorokieselsäure
More selective reagent than HF for the cleavage of silylgroups. Excess reagent is not required cf. HF and often catalytic amounts are sufficient. JOC., 57, 2492 (1992).
Formel:
H₂F₆Si Molecular Weight: 144,09 g/mol Schmelzpunkt: 15…17 °C Dichte: 1,22 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036289 CAS-Nummer: 16961-83-4 EINECS: 241-034-8 UN: 1778 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...