Pentafluoropropanal methyl hemiacetal
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pentafluoropropionaldehyde methyl hemiacetal, 1-Methoxy-1H-perfluoropropan-1-ol
PC4799-5G
APOSPC4799-5GEA
174
EUR
APOSPC4799-5G
Pentafluoropropanal methyl hemiacetal
Pentafluoropropanal methyl hemiacetal
Formel:
C₄H₅F₅O₂ Molecular Weight: 180,073 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039254 CAS-Nummer: 59872-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...