2-Nitro-4-(pentafluorosulphanyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3-nitrophenylsulphur pentafluoride

PC4815-1G PC4815-250MG
APOSPC4815-1GEA 973 EUR
APOSPC4815-1G APOSPC4815-250MG
2-Nitro-4-(pentafluorosulphanyl)aniline 95%
2-Nitro-4-(pentafluorosulphanyl)aniline
Formel: C₆H₅F₅N₂O₂S
Molecular Weight: 264.17 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11501034
CAS-Nummer: 197385-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR