2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine

Lieferant: Apollo Scientific
Achtung

PC4818-5G
APOSPC4818-5GEA 140 EUR
APOSPC4818-5G
2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine
2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine
Formel: C₇H₃Cl₂F₅N₂
Molecular Weight: 281,01 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03424472
CAS-Nummer: 111234-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR