1-(4-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC48295-1G
APOSPC48295-1GEA 147 EUR
APOSPC48295-1G
1-(4-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₀FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01569311
CAS-Nummer: 56617-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR