6-Fluor-3-(4-piperidinyl)-1,2-benzoxazol 97%

Lieferant: Apollo Scientific
Gefahr

PC48296-25G
APOSPC48296-25GEA 109 EUR
APOSPC48296-25G
6-Fluor-3-(4-piperidinyl)-1,2-benzoxazol 97%
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole
Formel: C₁₂H₁₃FN₂O
Lagertemperatur: Kühlschrank
MDL: MFCD03236724
CAS-Nummer: 84163-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR