2-Chlor-6-(trifluormethyl)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC48302-25G
PC48302-5G
PC48302-100G
APOSPC48302-25GEA
279
EUR
APOSPC48302-25G
APOSPC48302-5G
APOSPC48302-100G
2-Chlor-6-(trifluormethyl)benzylbromid 98%
2-Chlor-6-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrClF₃ Dichte: 1,6951 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660232 CAS-Nummer: 886500-26-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...