2-Iodo-5-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC48309-1G
PC48309-25G
PC48309-5G
APOSPC48309-1GEA
0
EUR
APOSPC48309-1G
APOSPC48309-25G
APOSPC48309-5G
2-Iodo-5-(trifluoromethyl)benzoic acid 97%
2-Iodo-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₄F₃IO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11849931 CAS-Nummer: 702641-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...