2-Iodo-5-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC48309-1G PC48309-25G PC48309-5G
APOSPC48309-1GEA 0 EUR
APOSPC48309-1G APOSPC48309-25G APOSPC48309-5G
2-Iodo-5-(trifluoromethyl)benzoic acid 97%
2-Iodo-5-(trifluoromethyl)benzoic acid
Formel: C₈H₄F₃IO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11849931
CAS-Nummer: 702641-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR