4-Amino-2-chloro-5-fluoropyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-fluoro-4-pyridinamine

PC48311-1G
APOSPC48311-1GEA 223 EUR
APOSPC48311-1G
4-Amino-2-chloro-5-fluoropyridine
4-Amino-2-chloro-5-fluoropyridine
Formel: C₅H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16622225
CAS-Nummer: 89510-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR