3,4-Difluor-o-toluylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Difluoro-2-methylbenzoic acid

PC48321-25G
APOSPC48321-25GEA 169 EUR
APOSPC48321-25G
3,4-Difluor-o-toluylsäure
3,4-Difluor-o-toluylsäure
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07777153
CAS-Nummer: 157652-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR