5-Brom-2-hydroxy-3-(trifluormethyl)pyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-(trifluoromethyl)pyridin-2-ol

PC48328-25G
APOSPC48328-25GEA 244 EUR
APOSPC48328-25G
5-Brom-2-hydroxy-3-(trifluormethyl)pyridin 98%
5-Brom-2-hydroxy-3-(trifluormethyl)pyridin
Formel: C₆H₃BrF₃NO
Molecular Weight: 242.00 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546486
CAS-Nummer: 76041-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR