4-Bromo-3-fluoroaniline, N-CBZ protected 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzyl (4-bromo-3-fluorophenyl)carbamate, Phenylmethyl (4-bromo-3-fluorophenyl)carbamate

PC48342-1G
APOSPC48342-1GEA 42.2 EUR
APOSPC48342-1G
4-Bromo-3-fluoroaniline, N-CBZ protected 97%
4-Bromo-3-fluoroaniline, N-CBZ protected
Formel: C₁₄H₁₁BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23381075
CAS-Nummer: 510729-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR