2-(Difluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC48345-1G
PC48345-250MG
PC48345-5G
APOSPC48345-1GEA
0
EUR
APOSPC48345-1G
APOSPC48345-250MG
APOSPC48345-5G
2-(Difluoromethyl)benzoic acid 97%
2-(Difluoromethyl)benzoic acid
Formel:
C₈H₆F₂O₂ Schmelzpunkt: 128…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140191 CAS-Nummer: 799814-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...