3'-Fluor-2'-methoxyacetophenon 97%

Lieferant: Apollo Scientific
Achtung

PC48354-1G PC48354-250MG PC48354-5G
APOSPC48354-1GEA 69.6 EUR
APOSPC48354-1G APOSPC48354-250MG APOSPC48354-5G
3'-Fluor-2'-methoxyacetophenon 97%
3'-Fluor-2'-methoxyacetophenon
Formel: C₉H₉FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12545933
CAS-Nummer: 295779-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR