2-(Difluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC48358-1G PC48358-250MG
APOSPC48358-1GEA 0 EUR
APOSPC48358-1G APOSPC48358-250MG
2-(Difluoromethyl)benzyl alcohol
2-(Difluoromethyl)benzyl alcohol
Formel: C₈H₈F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD26728124
CAS-Nummer: 1018678-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR