2-Chloro-6-(trifluoromethyl)pyrazine
Lieferant: Apollo Scientific
|
Gefahr
|
PC48361-10G
PC48361-1G
PC48361-5G
APOSPC48361-10GEA
1070
EUR
APOSPC48361-10G
APOSPC48361-1G
APOSPC48361-5G
2-Chloro-6-(trifluoromethyl)pyrazine
2-Chloro-6-(trifluoromethyl)pyrazine
Formel:
C₅H₂ClF₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189477 CAS-Nummer: 61655-69-4 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...