2-Chloro-5-(trifluoromethyl)pyrazine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC48362-1G
PC48362-5G
APOSPC48362-1GEA
0
EUR
APOSPC48362-1G
APOSPC48362-5G
2-Chloro-5-(trifluoromethyl)pyrazine 97%
2-Chloro-5-(trifluoromethyl)pyrazine
Formel:
C₅H₂ClF₃N₂ Siedepunkt: 85 °C/112mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189478 CAS-Nummer: 799557-87-2 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...