4-Nitro-2-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC48366-1G PC48366-5G
APOSPC48366-1GEA 0 EUR
APOSPC48366-1G APOSPC48366-5G
4-Nitro-2-(trifluoromethyl)benzyl bromide 97%
4-Nitro-2-(trifluoromethyl)benzyl bromide
Formel: C₈H₅BrF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11847129
CAS-Nummer: 694499-22-4
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR