Ethyl 6-bromo-7-fluorobenzo[b]furan-2-carboxylate, tech
Lieferant: Apollo Scientific
|
Achtung
|
PC48397-250MG
APOSPC48397-250MGEA
109
EUR
APOSPC48397-250MG
Ethyl 6-bromo-7-fluorobenzo[b]furan-2-carboxylate, tech
Ethyl 6-bromo-7-fluorobenzo[b]furan-2-carboxylate
Formel:
C₁₁H₈BrFO₃ Molecular Weight: 287.082 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31922514 CAS-Nummer: 2366994-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...