Methyl 4-methoxy-2-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC48410-1G PC48410-5G PC48410-25G
APOSPC48410-1GEA 0 EUR
APOSPC48410-1G APOSPC48410-5G APOSPC48410-25G
Methyl 4-methoxy-2-(trifluoromethyl)benzoate
Methyl 4-methoxy-2-(trifluoromethyl)benzoate
Formel: C₁₀H₉F₃O₃
Schmelzpunkt: 47…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06204092
CAS-Nummer: 773875-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR