Ethyl-2-chloro-5-fluoro-pyridine-3-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-chloro-5-fluoronicotinate

PC48412-1G
APOSPC48412-1GEA 52.2 EUR
APOSPC48412-1G
Ethyl-2-chloro-5-fluoro-pyridine-3-carboxylate 95%
Ethyl-2-chloro-5-fluoro-pyridine-3-carboxylate
Formel: C₈H₇ClFNO₂
Molecular Weight: 203,6 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05662395
CAS-Nummer: 139911-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR