2-Fluoro-5-(trifluoromethoxy)nitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
PC48423-1G
PC48423-5G
PC48423-25G
APOSPC48423-1GEA
0
EUR
APOSPC48423-1G
APOSPC48423-5G
APOSPC48423-25G
2-Fluoro-5-(trifluoromethoxy)nitrobenzene
2-Fluoro-5-(trifluoromethoxy)nitrobenzene
Formel:
C₇H₃F₄NO₃ Flammpunkt: >110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00042300 CAS-Nummer: 124170-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...