4-Brom-2-chlor-1-(trifluormethoxy)benzol 97%

Lieferant: Apollo Scientific
Achtung

PC48424-10G PC48424-1G PC48424-5G
APOSPC48424-10GEA 0 EUR
APOSPC48424-10G APOSPC48424-1G APOSPC48424-5G
4-Brom-2-chlor-1-(trifluormethoxy)benzol 97%
4-Brom-2-chlor-1-(trifluormethoxy)benzol
Formel: C₇H₃BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD10699114
CAS-Nummer: 158579-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR