3-Brom-4-(trifluormethoxy)benzonitril

Lieferant: Apollo Scientific
Achtung

PC48427-1G PC48427-5G
APOSPC48427-1GEA 219 EUR
APOSPC48427-1G APOSPC48427-5G
3-Brom-4-(trifluormethoxy)benzonitril
3-Brom-4-(trifluormethoxy)benzonitril
Formel: C₈H₃BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13195683
CAS-Nummer: 191602-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR