4-Bromo-3-chloro-2-fluorobenzaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC48449-1G
PC48449-250MG
PC48449-5G
APOSPC48449-1GEA
313
EUR
APOSPC48449-1G
APOSPC48449-250MG
APOSPC48449-5G
4-Bromo-3-chloro-2-fluorobenzaldehyde 97%
4-Bromo-3-chloro-2-fluorobenzaldehyde
Formel:
C₇H₃BrClFO Schmelzpunkt: 77…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD26958726 CAS-Nummer: 1696224-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...