6-Bromo-3-chloro-2-fluorobenzonitrile 95%

Lieferant: Apollo Scientific
Gefahr

PC48452-1G
APOSPC48452-1GEA 522 EUR
APOSPC48452-1G
6-Bromo-3-chloro-2-fluorobenzonitrile 95%
6-Bromo-3-chloro-2-fluorobenzonitrile
Formel: C₇H₂BrClFN
Molecular Weight: 234,45 g/mol
MDL: MFCD26958725
CAS-Nummer: 1779124-51-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR