6-Bromo-3-chloro-2-fluorophenacyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3'-Chloro-2,6'-dibromo-2'-fluoroacetophenone, 2-Bromo-1-(6-bromo-3-chloro-2-fluorophenyl)ethan-1-one

PC48480-1G PC48480-250MG
APOSPC48480-1GEA 383 EUR
APOSPC48480-1G APOSPC48480-250MG
6-Bromo-3-chloro-2-fluorophenacyl bromide
6-Bromo-3-chloro-2-fluorophenacyl bromide
Formel: C₈H₄Br₂ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD28096621
CAS-Nummer: 1804382-46-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR