2-Brom-1-iod-4-(trifluormethoxy)benzol

Lieferant: Apollo Scientific
Achtung

PC48517-25G PC48517-5G
APOSPC48517-25GEA 0 EUR
APOSPC48517-25G APOSPC48517-5G
2-Brom-1-iod-4-(trifluormethoxy)benzol
2-Brom-1-iod-4-(trifluormethoxy)benzol
Formel: C₇H₃BrF₃IO
Lagertemperatur: Kühlschrank
MDL: MFCD07369009
CAS-Nummer: 883546-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR