1-Bromo-4-iodo-2-(trifluoromethoxy)benzene 97%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-5-iodo-α,α,α-trifluoroanisole, 2-Bromo-5-iodophenyl trifluoromethyl ether

PC48518-250MG
APOSPC48518-250MGEA 72.5 EUR
APOSPC48518-250MG
1-Bromo-4-iodo-2-(trifluoromethoxy)benzene 97%
1-Bromo-4-iodo-2-(trifluoromethoxy)benzene
Formel: C₇H₃BrF₃IO
Molecular Weight: 366,902 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12761590
CAS-Nummer: 1187984-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR