3-Brom-4-iodbenzotrifluorid

Lieferant: Apollo Scientific
Achtung

PC48522-1G PC48522-25G PC48522-5G
APOSPC48522-1GEA 21.9 EUR
APOSPC48522-1G APOSPC48522-25G APOSPC48522-5G
3-Brom-4-iodbenzotrifluorid
3-Brom-4-iodbenzotrifluorid
Formel: C₇H₃BrF₃I
Lagertemperatur: Kühlschrank
MDL: MFCD08543938
CAS-Nummer: 481075-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR