3-Bromo-5-(trifluoromethyl)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-3-(bromomethyl)-5-(trifluoromethyl)benzene, 3-Bromo-5-(bromomethyl)benzotrifluoride
PC48526-5G
APOSPC48526-5GEA
263
EUR
APOSPC48526-5G
3-Bromo-5-(trifluoromethyl)benzyl bromide
3-Bromo-5-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅Br₂F₃ Molecular Weight: 317,929 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11044989 CAS-Nummer: 954123-46-7 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...