3-(4-Fluorophenyl)pentanedioic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Fluorophenyl)glutaric acid

PC48529-250MG
APOSPC48529-250MGEA 48 EUR
APOSPC48529-250MG
3-(4-Fluorophenyl)pentanedioic acid 95+%
3-(4-Fluorophenyl)pentanedioic acid
Formel: C₁₁H₁₁FO₄
Molecular Weight: 226,201 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19352347
CAS-Nummer: 3449-63-6
EINECS: 222-373-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR