(1R)-1-(3,4-Difluorophenyl)ethylamine hydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(3,4-Difluorophenyl)ethanamine hydrochloride, (R)-3,4-Difluoro-α-methylbenzylamine hydrochloride

PC48534-250MG
APOSPC48534-250MGEA 236 EUR
APOSPC48534-250MG
(1R)-1-(3,4-Difluorophenyl)ethylamine hydrochloride 95+%
(1R)-1-(3,4-Difluorophenyl)ethylamine hydrochloride
Formel: C₈H₉F₂N·ClH
Molecular Weight: 193,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11865391
CAS-Nummer: 441074-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR