(1R)-1-(3,5-Difluorophenyl)ethylamine hydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(3,5-Difluorophenyl)ethanamine hydrochloride, (R)-3,5-Difluoro-α-methylbenzylamine hydrochloride

PC48536-250MG
APOSPC48536-250MGEA 98.6 EUR
APOSPC48536-250MG
(1R)-1-(3,5-Difluorophenyl)ethylamine hydrochloride 95+%
(1R)-1-(3,5-Difluorophenyl)ethylamine hydrochloride
Formel: C₈H₉F₂N·ClH
Molecular Weight: 193,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101200
CAS-Nummer: 321318-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR