(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 95+%

Lieferant: Apollo Scientific

PC48541-250MG
APOSPC48541-250MGEA 111 EUR
APOSPC48541-250MG
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 95+%
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride
Formel: C₈H₁₀ClF₂N
Molecular Weight: 193,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101244
CAS-Nummer: 844647-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR