(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(3-Chloro-2-fluorophenyl)ethanamine hydrochloride

PC48549-250MG
APOSPC48549-250MGEA 37.7 EUR
APOSPC48549-250MG
(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride 95+%
(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride
Formel: C₈H₉ClFN·ClH
Molecular Weight: 210,076 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12758015
CAS-Nummer: 1253792-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR