4-Fluor-3-(4-morpholinylcarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-(morpholine-4-carbonyl)phenylboronic acid

PC4855-1G
APOSPC4855-1GEA 256 EUR
APOSPC4855-1G
4-Fluor-3-(4-morpholinylcarbonyl)phenylboronsäure 98%
4-Fluor-3-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₄
Schmelzpunkt: 138…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235048
CAS-Nummer: 874219-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR