(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(4-Chloro-3-fluorophenyl)ethanamine hydrochloride, (R)-4-Chloro-3-fluoro-α-methylbenzylamine hydrochloride

PC48555-100MG PC48555-250MG PC48555-1G
APOSPC48555-100MGEA 197 EUR
APOSPC48555-100MG APOSPC48555-250MG APOSPC48555-1G
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride 98%
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride
Formel: C₈H₉ClFN·ClH
Molecular Weight: 210.076 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12547495
CAS-Nummer: 1253790-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR