(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride 98%
Lieferant: Apollo Scientific
Synonyme:
(R)-1-(4-Chloro-3-fluorophenyl)ethanamine hydrochloride, (R)-4-Chloro-3-fluoro-α-methylbenzylamine hydrochloride
PC48555-100MG
PC48555-250MG
PC48555-1G
APOSPC48555-100MGEA
197
EUR
APOSPC48555-100MG
APOSPC48555-250MG
APOSPC48555-1G
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride 98%
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride
Formel:
C₈H₉ClFN·ClH Molecular Weight: 210.076 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547495 CAS-Nummer: 1253790-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...