3-Difluoromethyl-5-phenylpyrazole

Lieferant: Apollo Scientific

PC4856-10G
APOSPC4856-10GEA 894 EUR
APOSPC4856-10G
3-Difluoromethyl-5-phenylpyrazole
3-Difluoromethyl-5-phenylpyrazole
Formel: C₁₀H₈F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04039285
CAS-Nummer: 122980-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR